ContactUsLinkComponent
iFluor® 555-PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
iFluor® 555-PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-526
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 555-PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-526
Supplier:  AAT BIOQUEST INC
Supplier Number:  17042
CAS Number:  
Specifications
  • Size:
    25 nmoles
  • Cat. no.:
    77461-526
Specifications
About this item
This iFluor® 555-PEG12-dUTP conjugate has the same spectral properties to ChromaTide™ Alexa Fluor 555-dUTP.
  • PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
Caution: For research use only.