ContactUsLinkComponent
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7.5)*
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-514
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-514
Supplier:  AAT BIOQUEST INC
Supplier Number:  17033
CAS Number:  
Specifications
  • Size:
    25 nmoles
  • Cat. no.:
    77461-514
Specifications
About this item
iFluor® 560-dUTP is a iFluor 560-modified nucleotide that can be used in various molecular biology applications, such as labeling and detection of DNA molecules.
  • iFluor® 560-dUTP is preferred over other similar fluorescent dyes due to its high fluorescence intensity, photostability and compatibility with a wide range of detection systems
  • Used in techniques such as fluorescence in situ hybridization (FISH), microarray analysis and real-time PCR
When incorporated into DNA during synthesis, the iFluor® 560-dUTP can be visualized using fluorescence microscopy or other fluorescence-based assays.
Caution: For research use only.