CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing buffer is a buffer optimized for the multicolor flow cytometric analysis of cells.
- Designed for ease of use
- Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
: For research use only.
Frequently Bought Together