ContactUsLinkComponent
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # 77462-178
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # 77462-178
Supplier:  AAT Bioquest
Supplier Number:  31005
CAS Number:  
Specifications
  • Description:
    ReadiUse™ Universal kinase reaction stopping buffer, 100 ml
  • Cat. no.:
    77462-178
Specifications
About this item
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
  • Designed for ease of use
  • Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Caution: For research use only.