PBS Tablet (Phosphate Buffered Saline Tablet)
PBS Tablet (Phosphate Buffered Saline Tablet)
Phosphate buffered saline tablet, 10 tablets
Catalog # 77461-346
Supplier:   AAT Bioquest
Supplier #:  10010
PBS Tablet (Phosphate Buffered Saline Tablet)
Catalog # 77461-346
Supplier:  AAT Bioquest
CAS Number:  
PBS Tablet (Phosphate Buffered Saline Tablet)
PBS Tablet (Phosphate Buffered Saline Tablet)
Phosphate buffered saline tablet, 10 tablets
CAS Number:  
Price
$78.48
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
  • Designed for ease of use
  • Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Caution: For research use only.
Documents
Spécifications
Spécifications
  • Capacité:
    10 tablets
  • Désignation:
    Phosphate buffered saline tablet