ContactUsLinkComponent
Logic® Mag-Zone™ Magnetic Pipette Rack
Catalog # 77574-618
Supplier:  Labconco
New Product
undefined
Logic® Mag-Zone™ Magnetic Pipette Rack
Catalog # 77574-618
Supplier:  Labconco
Supplier Number:  3033000
New Product
Specifications
  • Description:
    Magnetic pipette rack holds 4 standard lab pipettes and one standard pipette aid, 2.3 kg
  • Cat. no.:
    77574-618
Specifications
About this item
Logic Mag-Zone magnetic pipette rack holds four standard lab pipettes and one standard pipette aid.
The Mag-Zone magnetic pipette rack allows for organization capabilities inside your cabinet. It holds four standard lab pipettes and one standard pipette aid. Engineered from stainless steel it can easily be decontaminated. This magnetic accessory is designed for use inside the Labconco Logic biosafety cabinet.