uPIPETTE™ Adjustable Volume Ergonomic Universal Pipettes, Single Channel
: Biotix
New Product
uPIPETTES™ Offer accuracy and precision and universal compatibility. Biotix uses technology to reduce fatigue and makes the uPIPETTE a top choice to improve your laboratory’s ergonomics.
- Compatible with Biotix uTIP and other universal pipette tips
- Low drag seals and light spring forces produce a smooth accurate draw with lighter force
- Easy to reach ejector button is cushioned to reduce dynamic stresses when ejecting a tip
- Volume lock ensures your measurements are accurate and precise without volume drift
- Designed to be rugged and durable for long term use with a low cost of ownership
- Unique serial ID number under the finger hook for GLP/GMP compliance
Pipetting is a highly repetitive motion which can have lasting effects on your hand, wrist and shoulder. Biotix® uPIPETTE™ is designed to improve the ergonomics of traditional pipetting systems by reducing the forces you’re exposed to during this repetitive process. uPIPETTE produce the low ejection forces, reducing your risk of developing repetitive stress injuries and muscle fatigue.
: ISO 8655 certified.
: Calibration certificate included in the box.
Frequently Bought Together