Storage Bins
Supplier:   Treston
Storage Bins
Supplier:  Treston
Certificates
Storage Bins
90 Options Available Below
Product Details & Documents
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects
 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
Product Family Options
ESD Picking Bin
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • ESD picking bins
    Black
    Case of 60
    6.3×3.9×2.8"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $957.61
    $957.61
    Case of 60
       $957.61
    Supplier Number: 30-1L-4ESD
ESD Stacking Bin Sets
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • ESD stacking bin set (Four SOVE1015-4ESD bins)
    Black
    PP
    Pack of 4
    6.5×4.13×2.95"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $53.92
    $53.92
    Pack of 1
       $53.92
    Supplier Number: SBS4-1015-4ESD
  • ESD stacking bin set (Two SOVE1525-6 and two SOVE1520-6)
    Black
    PP
    Pack of 4
    (2) 9.84×5.87×5.12" and (2) 7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $143.64
    $143.64
    Pack of 1
       $143.64
    Supplier Number: SBS4-2ESD
ESD Stacking Bins
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • ESD stacking bins
    Black
    PP
    Case of 60
    6.50×4.13×2.95"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $887.32
    $887.32
    Case of 60
       $887.32
    Supplier Number: 1015-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 45
    7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Case of 45
    Case of 45 - $1,557.58
    $1,557.58
    Case of 45
       $1,557.58
    Supplier Number: 1520-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 30
    9.84×5.87×5.12"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $1,514.95
    $1,514.95
    Case of 30
       $1,514.95
    Supplier Number: 1525-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 24
    11.80×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $2,099.72
    $2,099.72
    Case of 24
       $2,099.72
    Supplier Number: 1930-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 24
    15.74×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $2,562.56
    $2,562.56
    Case of 24
       $2,562.56
    Supplier Number: 1940-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 12
    19.70×7.32×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $2,272.96
    $2,272.96
    Case of 12
       $2,272.96
    Supplier Number: 1950-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 12
    15.60×12.2×6.14"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $1,617.78
    $1,617.78
    Case of 12
       $1,617.78
    Supplier Number: 3040-4ESD
  • ESD stacking bins
    Black
    PP
    Case of 8
    19.70×12.2×7.17"
    Log in for availability
    Log in for availability
    Case of 8
    Case of 8 - $2,525.74
    $2,525.74
    Case of 8
       $2,525.74
    Supplier Number: 3050-4ESD
Stacking Bins
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • Stacking bins
    Grey
    PP
    Case of 12
    19.70×7.32×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $933.49
    $933.49
    Case of 12
       $933.49
    Supplier Number: 1950-3
  • Stacking bins
    Blue
    PP
    Case of 45
    7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Case of 45
    Case of 45 - $945.06
    $945.06
    Case of 45
       $945.06
    Supplier Number: 1520-6
  • Stacking bins
    Blue
    PP
    Case of 24
    11.80×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,006.94
    $1,006.94
    Case of 24
       $1,006.94
    Supplier Number: 1930-6
  • Stacking bins
    Clear
    Polystyrene
    Case of 60
    6.50×4.13×2.95"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $640.69
    $640.69
    Case of 60
       $640.69
    Supplier Number: 1015-1
  • Stacking bins
    Red
    PP
    Case of 30
    9.84×5.87×5.12"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.75
    $898.75
    Case of 30
       $898.75
    Supplier Number: 1525-5
  • Stacking bins
    Blue
    PP
    Case of 30
    9.84×5.87×5.12"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.80
    $898.80
    Case of 30
       $898.80
    Supplier Number: 1525-6
  • Stacking bins
    Clear
    Polystyrene
    Case of 24
    11.80×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,019.89
    $1,019.89
    Case of 24
       $1,019.89
    Supplier Number: 1930-1
  • Stacking bins
    Grey
    PP
    Case of 24
    11.80×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $940.06
    $940.06
    Case of 24
       $940.06
    Supplier Number: 1930-3
  • Stacking bins
    Red
    PP
    Case of 24
    11.80×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $936.35
    $936.35
    Case of 24
       $936.35
    Supplier Number: 1930-5
  • Stacking bins
    Clear
    Polystyrene
    Case of 24
    15.74×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,299.21
    $1,299.21
    Case of 24
       $1,299.21
    Supplier Number: 1940-1
  • Stacking bins
    Grey
    PP
    Case of 24
    15.74×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,219.62
    $1,219.62
    Case of 24
       $1,219.62
    Supplier Number: 1940-3
  • Stacking bins
    Red
    PP
    Case of 24
    15.74×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,219.62
    $1,219.62
    Case of 24
       $1,219.62
    Supplier Number: 1940-5
  • Stacking bins
    Blue
    PP
    Case of 24
    15.74×7.32×6.14"
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $1,219.62
    $1,219.62
    Case of 24
       $1,219.62
    Supplier Number: 1940-6
  • Stacking bins
    Clear
    Polystyrene
    Case of 12
    19.70×7.32×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $975.14
    $975.14
    Case of 12
       $975.14
    Supplier Number: 1950-1
  • Stacking bins
    Red
    PP
    Case of 12
    19.70×7.32×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $933.49
    $933.49
    Case of 12
       $933.49
    Supplier Number: 1950-5
  • Stacking bins
    Blue
    PP
    Case of 12
    19.70×7.32×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $933.49
    $933.49
    Case of 12
       $933.49
    Supplier Number: 1950-6
  • Stacking bins
    Grey
    PP
    Case of 12
    15.60×12.2×6.14"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $790.69
    $790.69
    Case of 12
       $790.69
    Supplier Number: 3040-3
  • Stacking bins
    Red
    PP
    Case of 12
    15.60×12.2×6.14"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $790.69
    $790.69
    Case of 12
       $790.69
    Supplier Number: 3040-5
  • Stacking bins
    Blue
    PP
    Case of 12
    15.60×12.2×6.14"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $801.40
    $801.40
    Case of 12
       $801.40
    Supplier Number: 3040-6
  • Stacking bins
    Grey
    PP
    Case of 8
    19.70×12.2×7.17"
    Log in for availability
    Log in for availability
    Case of 8
    Case of 8 - $237.38
    $237.38
    Case of 8
       $237.38
    Supplier Number: 3050-3
  • Stacking bins
    Red
    PP
    Case of 8
    19.70×12.2×7.17"
    Log in for availability
    Log in for availability
    Case of 8
    Case of 8 - $1,042.14
    $1,042.14
    Case of 8
       $1,042.14
    Supplier Number: 3050-5
  • Stacking bins
    Blue
    PP
    Case of 12
    19.70×12.2×7.17"
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $1,042.14
    $1,042.14
    Case of 12
       $1,042.14
    Supplier Number: 3050-6
  • Stacking bins
    Grey
    PP
    Case of 60
    6.50×4.13×2.95"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $540.40
    $540.40
    Case of 60
       $540.40
    Supplier Number: 1015-3
  • Stacking bins
    Red
    PP
    Case of 60
    6.50×4.13×2.95"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $539.35
    $539.35
    Case of 60
       $539.35
    Supplier Number: 1015-5
  • Stacking bins
    Blue
    PP
    Case of 60
    6.50×4.13×2.95"
    Log in for availability
    Log in for availability
    Case of 60
    Case of 60 - $578.20
    $578.20
    Case of 60
       $578.20
    Supplier Number: 1015-6
  • Stacking bins
    Clear
    Polystyrene
    Case of 45
    7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Case of 45
    Case of 45 - $955.66
    $955.66
    Case of 45
       $955.66
    Supplier Number: 1520-1
  • Stacking bins
    Grey
    PP
    Case of 45
    7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Case of 45
    Case of 45 - $945.06
    $945.06
    Case of 45
       $945.06
    Supplier Number: 1520-3
  • Stacking bins
    Red
    PP
    Case of 45
    7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Case of 45
    Case of 45 - $1,011.22
    $1,011.22
    Case of 45
       $1,011.22
    Supplier Number: 1520-5
  • Stacking bins
    Clear
    Polystyrene
    Case of 30
    9.84×5.87×5.12"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $911.78
    $911.78
    Case of 30
       $911.78
    Supplier Number: 1525-1
  • Stacking bins
    Grey
    PP
    Case of 30
    9.84×5.87×5.12"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.80
    $898.80
    Case of 30
       $898.80
    Supplier Number: 1525-3
Shelf Bin Sets
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • Shelf bin set (Six SOVE3010-6 bins)
    Blue
    PP
    Pack of 6
    11.81×3.62×3.23"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $100.72
    $100.72
    Pack of 1
       $100.72
    Supplier Number: SBS6-3010-6
  • Shelf bin set (Six SOVE3015-6 bins)
    Blue
    PP
    Pack of 6
    11.81×5.20×3.94"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $138.18
    $138.18
    Pack of 1
       $138.18
    Supplier Number: SBS6-3015-6
  • Shelf bin set (Six SOVE3020-6 bins)
    Blue
    PP
    Pack of 6
    11.81×7.32×3.23"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $154.87
    $154.87
    Pack of 1
       $154.87
    Supplier Number: SBS6-3020-6
ESD Shelf Bins
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • ESD shelf bin set (Six SOVE3010-4ESD bins)
    Black
    Pack of 6
    3.62×11.81×3.23"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $180.46
    $180.46
    Pack of 1
       $180.46
    Supplier Number: SBS6-3010-4ESD
  • ESD Shelf bin set (Six SOVE3020-4ESD bins)
    Black
    Pack of 6
    7.32×11.81×3.23"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $246.47
    $246.47
    Pack of 1
       $246.47
    Supplier Number: SBS6-3020-4ESD
Shelf Bins
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • Shelf bins
    Blue
    PP
    Case of 20
    11.81×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $539.98
    $539.98
    Case of 20
       $539.98
    Supplier Number: 3015-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 30
    11.81×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $629.24
    $629.24
    Case of 30
       $629.24
    Supplier Number: 3010-1
  • Shelf bins
    Grey
    PP
    Case of 30
    11.81×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $630.05
    $630.05
    Case of 30
       $630.05
    Supplier Number: 3010-3
  • Shelf bins
    Red
    PP
    Case of 30
    11.81×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $630.05
    $630.05
    Case of 30
       $630.05
    Supplier Number: 3010-5
  • Shelf bins
    Blue
    PP
    Case of 30
    11.81×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $630.05
    $630.05
    Case of 30
       $630.05
    Supplier Number: 3010-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 30
    15.75×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $811.32
    $811.32
    Case of 30
       $811.32
    Supplier Number: 4010-1
  • Shelf bins
    Grey
    PP
    Case of 30
    15.75×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $718.16
    $718.16
    Case of 30
       $718.16
    Supplier Number: 4010-3
  • Shelf bins
    Red
    PP
    Case of 30
    15.75×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $718.16
    $718.16
    Case of 30
       $718.16
    Supplier Number: 4010-5
  • Shelf bins
    Blue
    PP
    Case of 30
    15.75×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $719.60
    $719.60
    Case of 30
       $719.60
    Supplier Number: 4010-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 30
    19.70×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.81
    $898.81
    Case of 30
       $898.81
    Supplier Number: 5010-1
  • Shelf bins
    Grey
    PP
    Case of 30
    19.70×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.76
    $898.76
    Case of 30
       $898.76
    Supplier Number: 5010-3
  • Shelf bins
    Red
    PP
    Case of 30
    19.70×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.76
    $898.76
    Case of 30
       $898.76
    Supplier Number: 5010-5
  • Shelf bins
    Blue
    PP
    Case of 30
    19.70×3.62×3.23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $898.76
    $898.76
    Case of 30
       $898.76
    Supplier Number: 5010-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 20
    11.81×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $538.99
    $538.99
    Case of 20
       $538.99
    Supplier Number: 3015-1
  • Shelf bins
    Grey
    PP
    Case of 20
    11.81×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $538.99
    $538.99
    Case of 20
       $538.99
    Supplier Number: 3015-3
  • Shelf bins
    Red
    PP
    Case of 20
    11.81×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $538.99
    $538.99
    Case of 20
       $538.99
    Supplier Number: 3015-5
  • Shelf bins
    Clear
    Polystyrene
    Case of 20
    15.75×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $657.96
    $657.96
    Case of 20
       $657.96
    Supplier Number: 4015-1
  • Shelf bins
    Grey
    PP
    Case of 20
    15.75×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $658.88
    $658.88
    Case of 20
       $658.88
    Supplier Number: 4015-3
  • Shelf bins
    Red
    PP
    Case of 20
    15.75×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $705.01
    $705.01
    Case of 20
       $705.01
    Supplier Number: 4015-5
  • Shelf bins
    Blue
    PP
    Case of 20
    15.75×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $658.88
    $658.88
    Case of 20
       $658.88
    Supplier Number: 4015-6
  • Shelf bins
    Grey
    PP
    Case of 20
    19.70×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $835.14
    $835.14
    Case of 20
       $835.14
    Supplier Number: 5015-3
  • Shelf bins
    Red
    PP
    Case of 20
    19.70×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $778.34
    $778.34
    Case of 20
       $778.34
    Supplier Number: 5015-5
  • Shelf bins
    Blue
    PP
    Case of 20
    19.70×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 20
    Case of 20 - $778.34
    $778.34
    Case of 20
       $778.34
    Supplier Number: 5015-6
  • Shelf bins
    Grey
    PP
    Case of 15
    23.62×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6015-3
  • Shelf bins
    Red
    PP
    Case of 15
    23.62×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6015-5
  • Shelf bins
    Blue
    PP
    Case of 15
    23.62×5.2×3.94"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6015-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 15
    11.81×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $481.25
    $481.25
    Case of 15
       $481.25
    Supplier Number: 3020-1
  • Shelf bins
    Grey
    PP
    Case of 15
    11.81×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $449.35
    $449.35
    Case of 15
       $449.35
    Supplier Number: 3020-3
  • Shelf bins
    Red
    PP
    Case of 15
    11.81×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $480.81
    $480.81
    Case of 15
       $480.81
    Supplier Number: 3020-5
  • Shelf bins
    Blue
    PP
    Case of 15
    11.81×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $452.40
    $452.40
    Case of 15
       $452.40
    Supplier Number: 3020-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 15
    15.75×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $539.46
    $539.46
    Case of 15
       $539.46
    Supplier Number: 4020-1
  • Shelf bins
    Grey
    PP
    Case of 15
    15.75×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $494.17
    $494.17
    Case of 15
       $494.17
    Supplier Number: 4020-3
  • Shelf bins
    Red
    PP
    Case of 15
    15.75×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $528.75
    $528.75
    Case of 15
       $528.75
    Supplier Number: 4020-5
  • Shelf bins
    Blue
    PP
    Case of 15
    15.75×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $494.17
    $494.17
    Case of 15
       $494.17
    Supplier Number: 4020-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 15
    19.69×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $630.43
    $630.43
    Case of 15
       $630.43
    Supplier Number: 5020-1
  • Shelf bins
    Grey
    PP
    Case of 15
    19.69×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $628.95
    $628.95
    Case of 15
       $628.95
    Supplier Number: 5020-3
  • Shelf bins
    Red
    PP
    Case of 15
    19.69×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $628.95
    $628.95
    Case of 15
       $628.95
    Supplier Number: 5020-5
  • Shelf bins
    Blue
    PP
    Case of 15
    19.69×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $673.00
    $673.00
    Case of 15
       $673.00
    Supplier Number: 5020-6
  • Shelf bins
    Clear
    Polystyrene
    Case of 15
    23.62×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $719.71
    $719.71
    Case of 15
       $719.71
    Supplier Number: 6020-1
  • Shelf bins
    Grey
    PP
    Case of 15
    23.62×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6020-3
  • Shelf bins
    Red
    PP
    Case of 15
    23.62×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6020-5
  • Shelf bins
    Blue
    PP
    Case of 15
    23.62×7.32×3.23"
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $675.16
    $675.16
    Case of 15
       $675.16
    Supplier Number: 6020-6
Stacking Bin Sets
Product Information
  • Désignation
    Color
    Matière
    Taille
    Dimensions
    Availability
    Price
  • Stacking bin set (Four SOVE1015-6 bins)
    Blue
    PP
    Pack of 4
    6.5×4.13×2.95"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $31.94
    $31.94
    Pack of 1
       $31.94
    Supplier Number: SBS4-1015-6
  • Stacking bin set (Two SOVE1525-6 and two SOVE1520-6)
    Blue
    PP
    Pack of 4
    (2) 9.84×5.87×5.12" and (2) 7.56×5.87×4.13"
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $85.17
    $85.17
    Pack of 1
       $85.17
    Supplier Number: SBS4-2