Logic® Washable Prefilter Kits | Avantor
Logic® Washable Prefilter Kits
Supplier:   Labconco
Logic® Washable Prefilter Kits
Supplier:  Labconco
Certificates
Logic® Washable Prefilter Kits
Logic® Washable Prefilter Kits
4 Options Available Below
Product Details & Documents
Washable prefilter kit for Logic biosafety cabinets.
  • Washable metal screen pre-filter
  • Installation hardware included
  • Compatible with Legacy Purifier Logic
  • For use with A2 and B2 biosafety cabinets
Washable Prefilters clean incoming air of dirt, dust and particles extending the life of the HEPA filter and blower motors. Each kit consists of a washable/reusable, metal screen-type prefilter and installation hardware. Legacy Purifier Logic compatible. The washable prefilter kits are designed for A2 and B2 biosafety cabinets only. Installation is required.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Washable prefilter kit
    92.1×73.7×15.7 cm (36.25W×29D×6.2"H)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.00
    $356.00
    Each
       $356.00
    Supplier Number: 3288600
  • Washable prefilter kit
    122.6×73.7×15.7 cm (48.25W×29D×6.2"H)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $427.00
    $427.00
    Each
       $427.00
    Supplier Number: 3288601
  • Washable prefilter kit
    153.0×73.7×15.7 cm (60.25W×29D×6.2"H)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $490.00
    $490.00
    Each
       $490.00
    Supplier Number: 3288602
  • Washable prefilter kit
    183.5×73.7×15.7 cm (72.25W×29D×6.2"H)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $567.00
    $567.00
    Each
       $567.00
    Supplier Number: 3288603