À propos de cet article
Normal feline (cat) serum, unconjugated and lyophilized.
Reconstitution: Rehydrate with 11.0 ml of deionized water. Swirl gently and let stand for up to 2 hours at 18 to 25 °C. Centrifuge reconstituted serum to remove any precipitates.
Storage: Store lyophilized material at 2 to 8 °C. For long term storage after reconstitution, prepare small aliquots and store at −20 °C . For storage at 2 to 8 °C, add a preservative to prevent growth of bacteria.
Cross Reactivity: Serum may may contain anti-feline calicivirus antibodies. Not suitable as negative control within calicivirus assay.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77553-606
- 77553-608
- Concentration:
- 60 mg/ml
- 60 mg/ml
- Purification:
- Lipid extracted and dialyzed against 10 mM sodium phosphate, 0.15 M sodium chloride
- Lipid extracted and dialyzed against 10 mM sodium phosphate, 0.15 M sodium chloride
- ELISA:
- Yes
- Yes
- Western blot:
- Yes
- Yes
- Storage buffer:
- 10 mM sodium phosphate, 0.15 M sodium chloride, pH 7.2
- 10 mM sodium phosphate, 0.15 M sodium chloride, pH 7.2
- Storage temperature:
- −20 °C
- −20 °C