Burn Dressing
Burn dressing, 19.84 lbs., 2×6"
Catalog # 77565-682
Supplier: Dukal
Fournisseur #: 7007101
Jump to:
Product Details & Documents
Soothes and provides cooling to burn.
- Non-adherent
- Not made with natural latex
- Sterile
This dressing helps prevent wound contamination.
Packaging: 1/pack, 12 packs/box, 16 boxes/case
Spécifications
- Désignation:Burn dressing, 19.84 lbs.
- Taille:2×6"