General Thromboxane A2 (TXA2) ELISA Kit
48, 1 kit
Catalog # 77538-420
Supplier: Bioss
Supplier #: BSKL61349-48T
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results
Spécifications
Spécifications
- Taille:1 kit
- No. of Tests:48
- Cat. No.:77538-420
- Assay duration:Multiple steps
- Detection method:Colorimetric microplate reader primary wavelength 450 nm
- Sensitivity:5.62 pg/ml
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Species independent
- Target protein:Thromboxane A2 (TXA2)
- Detection range:15.6 - 1000 pg/ml
- Storage temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Shelf life:12 Months
- Assay Type:Competitive
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



