Supertek® Resonance Apparatus
470356-598
New Product
- Description:Resonance apparatus
- Cat. no.:470356-598
An attractive an easy to use resonance apparatus.
- Tuning forks available separately
Investigate the length, frequency and velocity of sound waves by studying the resonance of air columns. A vibrating tuning fork, held over the top of the graduated water cylinder, will begin to resonate with the air column above the water in the cylinder. After taking the measurements of the air column and the tuning fork, the speed of sound can be calculated.