Provide high-quality, cost-effective options for large volume filtration needs. Systems are offered in multiple configurations and in sizes from 150 ml up to 1000 ml. Bottle top filter units fit Diamond® PureFlow™ solution bottles or other laboratory media bottles with GL45 threads.
- Sterile and ready for use
- Simple and efficient
- Versatile to meet most application needs
- Individually wrapped
- Irradiation sterile to SAL 10⁻⁶
- RNase and DNase free
- Non-pyrogenic
- Membrane and porosity are clearly marked on filter units
- Mult-step hose barb makes connections simple
- Angled hose barb keeps system stable on the benchtop
Low protein binding Polyether Sulfone (PES) membranes make Diamond PureFlow™ vacuum filters perfect for clarification, separation, or sterilization of aqueous solutions including cell culture media, biological samples, and laboratory reagents. Filter membranes are available in 0.10, 0.22 and 0.45 µl porosities to meet the requirements of most applications. Membrane and porosity are clearly marked on each filter unit.
Engineered with laboratory efficiency and safety in mind, Diamond® PureFlow™ vacuum filter systems include a multi-step, angled hose barb that accepts multiple sizes of vacuum lines while keeping the system stable on the benchtop. Graduation markings are included on both the filter unit and solution bottle for easy reference during and after filtration operations. Diamond® PureFlow™ vacuum filter systems are molded from virgin USP Class VI polystyrene (PS) and acrylonitrile butadiene styrene (ABS).
Performance
• High flow rates and low protein binding PES membranes for minimal sample loss
• Molded from virgin USP class VI resins
• Produced in class 100000 clean room