Facial Tissues
Supplier: Dukal
Jump to:
À propos de cet article
Facial tissues made of 100% virgin wood pulp.
- Fragrance free and without lotion
- Features colorful designs
- Available in standard and premium options
Packaging: Packaged in flat dispenser box made from recycled paper.
3 Options Available Below
Product Details & Documents
Facial tissues made of 100% virgin wood pulp.
- Fragrance free and without lotion
- Features colorful designs
- Available in standard and premium options
Packaging: Packaged in flat dispenser box made from recycled paper.
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationColorPlyDimensionsWeightPackagingAvailabilityPrice
- Facial tissues, standard, non sterileWhite28×8.3"9.9208 lbs.90/box, 30 boxes/caseLog in pour la disponibilitéCase of 2,700$43.48Supplier Number: TIS101
- Facial tissues, standard, non sterileWhite25.7×7"19.84 lbs.40/box, 200 boxes/caseLog in pour la disponibilitéCase of 8,000$107.49Supplier Number: TIS100
- Facial tissues, premium, non sterileWhite28×8.3"13.22774 lbs.100/box, 36 boxes/caseLog in pour la disponibilitéCase of 3,600$56.96Supplier Number: TIS102