Brady® Aggressive Adhesive Multi-Purpose Nylon Labels
Nylon, Black on white, BMP51;BMP53;M511, 1.5", —, —, 1.25", 160 labels/roll
Catalog # 76795-714
Supplier: Brady Worldwide
Supplier #: 170934
Brady® Aggressive Adhesive Multi-Purpose Nylon Labels
Catalog # 76795-714
Supplier: Brady Worldwide
Jump to:
Product Details & Documents
Versatile labels are ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates, and terminal blocks.
- Permanent acrylic adhesive
- Matte finish
- Autoclavable material; suitable for cryogenic storage
- Halogen-free
- Recommended ribbon: R4300 (CA94000-380)
Durable labels are designed to withstand extreme temperatures, chemicals, greases, oils, cleaners, and detergents. Permanent adhesive labels stick to smooth, rough, and powder-coated surfaces alike.
Certifications: UL recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).
Specifications
- Pour l'utilisation avec:BMP51;BMP53;M511
- Color:Black on white
- Hauteur:1.25"
- Longueur:—
- Matière:Nylon
- Packaging:160 labels/roll
- Width:1.5"
- Diameter:—