Brady® Self-Laminating Cryogenic Polyester Labels
Brady® Self-Laminating Cryogenic Polyester Labels
Supplier:   Brady Worldwide
Brady® Self-Laminating Cryogenic Polyester Labels
Supplier:  Brady Worldwide
Certificates
Brady® Self-Laminating Cryogenic Polyester Labels
Brady® Self-Laminating Cryogenic Polyester Labels
3 Options Available Below
Product Details & Documents
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides.
  • Permanent acrylic adhesive
  • Matte finish
  • Autoclavable, optically clear material is suitable for cryogenic storage
  • Recommended ribbon: R4300 (CA94000-380)
Dependable material is able to withstand extreme temperatures from −196 to +130 °C (−320 to +266 °F), immersion in liquid nitrogen, and autoclave use. Labels are resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% clorox solution. Made with a clear overlaminate that protects the print and maintains content visibility.
Product Family Options
Product Information
  • Matière
    Color
    Pour l'utilisation avec
    Dimensions
    Packaging
    Availability
    Price
  • Polyester
    Black on white/clear
    BMP41;BMP51;BMP53;M511
    0.6H×1.8"W
    180 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $115.25
    $115.25
    Each
       $115.25
    Supplier Number: 170769
  • Polyester
    Black on white
    BMP41;BMP51;BMP53;M511
    0.5"W × 25'L
    Log in for availability
    Log in for availability
    Each
    Each - $125.91
    $125.91
    Each
       $125.91
    Supplier Number: 170765
  • Polyester
    Black on white/clear
    BMP51;BMP53;M511
    1H × 1.75"W
    180 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $115.25
    $115.25
    Each
       $115.25
    Supplier Number: 170878