Double Rod Mini H-Stand Set | Avantor
Double Rod Mini H-Stand Set
Double rod mini H-stand set
Catalog # 77574-954
Fournisseur #:  A00000484
Double Rod Mini H-Stand Set
Catalog # 77574-954
Double Rod Mini H-Stand Set
Double Rod Mini H-Stand Set
Double rod mini H-stand set
Price
$1,853.17
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
The Mini H-stand set is a complete system with a base, double rod, clamp and a vessel holder with safety belt. It is designed to ensure maximum stability, precision and ease of positioning of the sample to be processed.
  • Enhanced stability and reduced vibrations through robust rods
  • Precision-engineered ergonomic knobs minimise operator fatigue and ensure consistent tightening
  • Efficient height adjustment is facilitated by an innovative easy-sliding technology system, enabling swift and precise adjustments
  • Reliability and simplified maintenance are ensured by the vessel holder with safety belt, providing steadfast support and easier cleaning procedures
The double rod mini H-stand set is the ideal solution for all applications that require absolute stability and precision.
Thanks to the double vertical rod configuration and the 'easy sliding' technology, the mini H-stand's sliding mechanism enables smooth and effortless movement for the operator.
In addition, it allows comfortable positioning of a 2.5 beaker without the need to tilt it, simply use the washable, rubber safety belt which provides maximum resistance to chemicals.
Spécifications
  • Désignation:
    Double rod mini H-stand set