SteriWare® Tablet Counting Triangles | Avantor
SteriWare® Tablet Counting Triangles
New Product
SteriWare® Tablet Counting Triangles
New Product
Certificates
SteriWare® Tablet Counting Triangles
SteriWare® Tablet Counting Triangles
2 Options Available Below
Product Details & Documents
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Packaging: Individually bagged in a heat-sealed laminate bag.
Product Family Options
Informations sur le produit
  • Désignation
    W×D×H
    Color
    Availability
    Price
  • Non-sterile
    230×230×13 mm
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $261.57
    $261.57
    Case of 30
       $261.57
    Supplier Number: 8066A-1
    Spécifications:
    New Product
    New Product
  • Sterile
    230×230×13 mm
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $347.99
    $347.99
    Case of 30
       $347.99
    Supplier Number: 8066A-1S
    Spécifications:
    New Product
    New Product