Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SteriWare® Trays

SteriWare® Trays

Sterile, 3000 ml, 265×310×55 mm
Catalog # 78116-998
New Product
Supplier #:  8081P-3000S
SteriWare® Trays
Catalog # 78116-998
New Product
SteriWare® Trays
SteriWare® Trays

Sterile, 3000 ml, 265×310×55 mm

Price
$607.67
Log in pour la disponibilité
Case of 30
Product Details & Documents

Trays made of PP, translucent.

  • Assembled and packed in a cleanroom
  • Individually bagged
  • FDA & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available sterile (gamma irradiated)

The disposable tray is ideal for hygienic applications within hospitals and pharmaceutical factories.

Spécifications
  • Capacité:
    3000 ml
  • Désignation:
    Sterile
  • W×D×H:
    265×310×55 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.