SteriWare® Sampling Spoons
SteriWare® Sampling Spoons
New Product
SteriWare® Sampling Spoons
New Product
Certificates
SteriWare® Sampling Spoons
SteriWare® Sampling Spoons
8 Options Available Below
Product Details & Documents
SteriWare® Single use long handled spoons are ideal for reaching into large containers or difficult to reach places. It can be used straight from the bag and manufactured under cleanroom conditions so they can be used without costly cleaning.
  • Made from FDA Conforming HDPE
  • Pre-sterilised versions are available
  • Moulded and suitable for powders and liquids
  • Fully validated sterilisation procedure (SAL 10⁻⁶)
  • Full batch traceability
Certifications: FDA, EU 10/2011 and EC 1935/2004 conformity; BSE/TSE free.
Packaging: Individually bagged in a heat-sealed laminate bag.
Product Family Options
Straight spoons
Product Information
  • Désignation
    Longueur
    Capacité
    Availability
    Price
  • Sampling spoons, non sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $382.11
    $382.11
    Case of 100
       $382.11
    Supplier Number: 8209H-10
    Specifications:
    New Product
    New Product
  • Sampling spoons, sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $483.48
    $483.48
    Case of 100
       $483.48
    Supplier Number: 8209H-10S
    Specifications:
    New Product
    New Product
  • Sampling spoons, non sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $385.23
    $385.23
    Case of 100
       $385.23
    Supplier Number: 8209H-20
    Specifications:
    New Product
    New Product
  • Sampling spoons, sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $486.60
    $486.60
    Case of 100
       $486.60
    Supplier Number: 8209H-20S
    Specifications:
    New Product
    New Product
Offset spoons
Product Information
  • Désignation
    Longueur
    Capacité
    Availability
    Price
  • Sampling spoons, non sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $417.59
    $417.59
    Case of 100
       $417.59
    Supplier Number: 8293H-20
    Specifications:
    New Product
    New Product
  • Sampling spoons, sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $488.08
    $488.08
    Case of 100
       $488.08
    Supplier Number: 8293H-20S
    Specifications:
    New Product
    New Product
  • Sampling spoons, non sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $414.21
    $414.21
    Case of 100
       $414.21
    Supplier Number: 8293H-10
    Specifications:
    New Product
    New Product
  • Sampling spoons, sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $499.65
    $499.65
    Case of 100
       $499.65
    Supplier Number: 8293H-10S
    Specifications:
    New Product