SteriWare® Dosing Spoons, PS | Avantor
SteriWare® Dosing Spoons, PS
New Product
SteriWare® Dosing Spoons, PS
New Product
Certificates
SteriWare® Dosing Spoons, PS
SteriWare® Dosing Spoons, PS
2 Options Available Below
Product Details & Documents
These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.
  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free
Packaging: Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • Non sterile
    2.5/5.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 130
    Case of 130 - $73.16
    $73.16
    Case of 130
       $73.16
    Supplier Number: 8642A-01
    Spécifications:
    New Product
    New Product
  • Sterile
    2.5/5.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 130
    Case of 130 - $102.07
    $102.07
    Case of 130
       $102.07
    Supplier Number: 8642A-01S
    Spécifications:
    New Product