SteriWare® Disposable Forceps
New Product
SteriWare® Disposable Forceps
New Product
Certificates
SteriWare® Disposable Forceps
SteriWare® Disposable Forceps
2 Options Available Below
Product Details & Documents
These diposable tweezers made from 100% virgin HDPE are ideal for picking up items up to 25 mm. Its special dual action design means it can pick up both small and large items.
  • Made from FDA and EC conforming material
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • Gamma irradiated (SAL 10⁻⁶)
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
 : Manufactured and packed in a class 100000 cleanroom environment. Individually bagged in a heat sealed PE bag.
Product Family Options
Informations sur le produit
  • Color
    Longueur
    Availability
    Price
  • White
    115 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $149.96
    $149.96
    Case of 100
       $149.96
    Supplier Number: 8025H-115
    Spécifications:
    New Product
    New Product
  • White
    115 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $183.44
    $183.44
    Case of 100
       $183.44
    Supplier Number: 8025H-115S
    Spécifications:
    New Product
    New Product