SteriWare® Jugs, PP | Avantor
SteriWare® Jugs, PP
New Product
SteriWare® Jugs, PP
New Product
Certificates
SteriWare® Jugs, PP
SteriWare® Jugs, PP
3 Options Available Below
Product Details & Documents
Made from PP (FDA, EC 1935/2004 and EU 10/2011 conforming).
  • Moulded and packed in a cleanroom
  • BSE/TSE free
  • Full batch traceability
  • Graduation marks at every 10 ml
These range of class leading jugs are available pre-sterilized (gamma irradiated). The ergonomic handle is rounded to make it comfortable to hold and use.
Packaging: Individually bagged.
Product Family Options
Informations sur le produit
  • Capacité
    Hauteur
    Sterility
    Ø
    Availability
    Price
  • 1000 ml
    170 mm
    Non sterile
    120 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $162.68
    $162.68
    Case of 18
       $162.68
    Supplier Number: 8432P-1000
    Spécifications:
    New Product
    New Product
  • 1000 ml
    170 mm
    Sterile
    120 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $300.60
    $300.60
    Case of 18
       $300.60
    Supplier Number: 8432P-1000S
    Spécifications:
    New Product
    New Product
  • 500 ml
    130 mm
    Non sterile
    90 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 35
    Case of 35 - $229.86
    $229.86
    Case of 35
       $229.86
    Supplier Number: 8432P-500
    Spécifications:
    New Product