Accupod Solid Samplers
Accupod Solid Samplers
New Product
Accupod Solid Samplers
New Product
Certificates
Accupod Solid Samplers
Accupod Solid Samplers
4 Options Available Below
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
  • Suitable for sampling blend uniformity
  • Next generation in sampling technology
  • Designed to minimize sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Volume
    Availability
    Price
  • Accupod sampler, non sterile
    HDPE
    1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $274.74
    $274.74
    Case of 50
       $274.74
    Supplier Number: 8360T-10
    Spécifications:
    New Product
    New Product
  • Accupod sampler, sterile
    HDPE
    1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $298.17
    $298.17
    Case of 50
       $298.17
    Supplier Number: 8360T-10S
    Spécifications:
    New Product
    New Product
  • Accupod sampler, non sterile
    HDPE
    5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $274.74
    $274.74
    Case of 50
       $274.74
    Supplier Number: 8360T-50
    Spécifications:
    New Product
    New Product
  • Accupod sampler, sterile
    HDPE
    5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $298.17
    $298.17
    Case of 50
       $298.17
    Supplier Number: 8360T-50S
    Spécifications:
    New Product