SteriWare® Capsule Counting Trays
New Product
SteriWare® Capsule Counting Trays
New Product
SteriWare® Capsule Counting Trays
SteriWare® Capsule Counting Trays
The disposable counting trays are made from robust, shatterproof PS. They are suitable for use with all capsules from size 2 to 00.
  • BSE/TSE free
  • Suitable for capsules size 2 to size 00
  • Manufactured and packed in a cleanroom
  • Can be used straight from the bag
 : Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
 : Individually bagged in a heat-sealed laminate bag.
Product Family Options
  • Sterility
    W×D×H
    Color
  • Non sterile
    260×130×10 mm
    White
    Case of 20
    Case of 20 - $169.97
    $169.97
    Case of 20
       $169.97
     : 8444A-1
    New Product
    New Product
  • Sterile
    260×130×10 mm
    White
    Case of 20
    Case of 20 - $256.78
    $256.78
    Case of 20
       $256.78
     : 8444A-1S