SteriWare® Spatulas, Micro, Disposable
New Product
Supplier: SAMPLING SYSTEMS LTD
Jump to:
À propos de cet article
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
- Moulded and packed in a cleanroom
- Suitable for food contact
- Available sterile (gamma irradiated)
- Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.
9 Options Available Below
Product Details & Documents
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
- Moulded and packed in a cleanroom
- Suitable for food contact
- Available sterile (gamma irradiated)
- Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.
Product Family Options
Informations sur le produit
- DésignationColorLongueurBlade LengthCapacitéBlade WidthSterilityAvailabilityPrice
- Supplier Number: 8015A-170Spécifications:New ProductNew Product
- Supplier Number: 8015A-170SSpécifications:New ProductNew Product
- Supplier Number: 8015H-170Spécifications:New ProductNew Product
- Supplier Number: 8015H-170SSpécifications:New ProductNew Product
- Supplier Number: 8015N-170Spécifications:New ProductNew Product
- Supplier Number: 8015N-170SSpécifications:New ProductNew Product
- Supplier Number: 8015V-170SSpécifications:New ProductNew Product
- Supplier Number: 8015NR-170SSpécifications:New ProductNew Product
- Supplier Number: 8015V-170Spécifications:New ProductNew Product