Axygen® LTS Compatible Pipet Tips
 :   Corning
Axygen® LTS Compatible Pipet Tips
 :  Corning
Axygen® LTS Compatible Pipet Tips
Axygen® LTS Compatible Pipet Tips
The new Axygen® LTS compatible pipet tips are ergonomically designed to reduce discomfort and are optimized to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting.
 : ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
  • Catalog No:
  • 77395-566
  • 77395-568
  • 77395-570
  • 77395-572
  • 77395-574
  • 77395-576
  • 77395-578
  • 77395-580
  • 77395-582
  • 77395-584
  • 77395-586
  • 77395-588
  • 77395-592
  • 77395-596
  • 77395-590
  • 77395-594
  • 77395-598
  • 77395-600
  • 77395-602
  • Environmentally Preferable:
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Environmental certification:
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
  • Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
Product Family Options
Racked, filtered, sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    +
    +
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $213.00
    $213.00
    Case of 3840
    Case of 3840 - $852.00
    $852.00
    Pack of 960
       $213.00
    Case of 3840
       $852.00
     : LTF-20-R-S
    Specifications
    Cat. No.
    77395-566
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    +
    +
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $170.00
    $170.00
    Case of 4800
    Case of 4800 - $850.00
    $850.00
    Pack of 960
       $170.00
    Case of 4800
       $850.00
     : LTF-200-R-S
    Specifications
    Cat. No.
    77395-568
    Environmentally Preferable
    Environmental certification
  • LTS compatible, extended length
    1000 µl
    +
    +
    98.58 mm
    Clear
    8 racks, 96 each
    Pack of 768
    Pack of 768 - $170.00
    $170.00
    Case of 3840
    Case of 3840 - $850.00
    $850.00
    Pack of 768
       $170.00
    Case of 3840
       $850.00
     : LTF-1000-R-S
    Specifications
    Cat. No.
    77395-570
    Environmentally Preferable
    Environmental certification
Racked, non-filtered, sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    +
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $94.90
    $94.90
    Case of 4800
    Case of 4800 - $474.50
    $474.50
    Pack of 960
       $94.90
    Case of 4800
       $474.50
     : LT-20-R-S
    Specifications
    Cat. No.
    77395-572
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    +
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $107.00
    $107.00
    Case of 4800
    Case of 4800 - $535.00
    $535.00
    Pack of 960
       $107.00
    Case of 4800
       $535.00
     : LT-200-R-S
    Specifications
    Cat. No.
    77395-574
    Environmentally Preferable
    Environmental certification
  • LTS compatible, extended length
    1000 µl
    +
    98.58 mm
    Clear
    8 racks, 96 each
    Pack of 768
    Pack of 768 - $108.00
    $108.00
    Case of 3840
    Case of 3840 - $540.00
    $540.00
    Pack of 768
       $108.00
    Case of 3840
       $540.00
     : LT-1000-R-S
    Specifications
    Cat. No.
    77395-576
    Environmentally Preferable
    Environmental certification
Racked, non-filtered, non sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $105.00
    $105.00
    Case of 4800
    Case of 4800 - $525.00
    $525.00
    Pack of 960
       $105.00
    Case of 4800
       $525.00
     : LT-20-R
    Specifications
    Cat. No.
    77395-578
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $103.00
    $103.00
    Case of 4800
    Case of 4800 - $515.00
    $515.00
    Pack of 960
       $103.00
    Case of 4800
       $515.00
     : LT-200-R
    Specifications
    Cat. No.
    77395-580
    Environmentally Preferable
    Environmental certification
  • LTS compatible, extended length
    1000 µl
    98.58 mm
    Clear
    8 racks, 96 each
    Pack of 768
    Pack of 768 - $103.00
    $103.00
    Case of 3840
    Case of 3840 - $515.00
    $515.00
    Pack of 768
       $103.00
    Case of 3840
       $515.00
     : LT-1000-R
    Specifications
    Cat. No.
    77395-582
    Environmentally Preferable
    Environmental certification
Reload, non-filtered, non sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $58.39
    $58.39
    Case of 3840
    Case of 3840 - $233.54
    $233.54
    Pack of 960
       $58.39
    Case of 3840
       $233.54
     : LT-20-RL
    Specifications
    Cat. No.
    77395-584
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $91.60
    $91.60
    Case of 3840
    Case of 3840 - $366.40
    $366.40
    Pack of 960
       $91.60
    Case of 3840
       $366.40
     : LT-200-RL
    Specifications
    Cat. No.
    77395-586
    Environmentally Preferable
    Environmental certification
  • LTS compatible, extended length
    1000 µl
    98.58 mm
    Clear
    8 racks, 96 each
    Pack of 768
    Pack of 768 - $90.70
    $90.70
    Case of 3072
    Case of 3072 - $362.80
    $362.80
    Pack of 768
       $90.70
    Case of 3072
       $362.80
     : LT-1000-RL
    Specifications
    Cat. No.
    77395-588
    Environmentally Preferable
    Environmental certification
Tip station stacked rack, non-filtered, sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    +
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $138.00
    $138.00
    Case of 4800
    Case of 4800 - $690.00
    $690.00
    Pack of 960
       $138.00
    Case of 4800
       $690.00
     : LT-20-TS-S
    Specifications
    Cat. No.
    77395-592
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    +
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $138.00
    $138.00
    Case of 4800
    Case of 4800 - $690.00
    $690.00
    Pack of 960
       $138.00
    Case of 4800
       $690.00
     : LT-200-TS-S
    Specifications
    Cat. No.
    77395-596
    Environmentally Preferable
    Environmental certification
Tip station stacked rack, non-filtered, non sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $131.00
    $131.00
    Case of 4800
    Case of 4800 - $655.00
    $655.00
    Pack of 960
       $131.00
    Case of 4800
       $655.00
     : LT-20-TS
    Specifications
    Cat. No.
    77395-590
    Environmentally Preferable
    Environmental certification
  • LTS compatible
    200 µl
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $130.00
    $130.00
    Case of 4800
    Case of 4800 - $650.00
    $650.00
    Pack of 960
       $130.00
    Case of 4800
       $650.00
     : LT-200-TS
    Specifications
    Cat. No.
    77395-594
    Environmentally Preferable
    Environmental certification
HybridRack, filtered, sterile
  • Tip Style
    Volume
    Sterile
    Filter
    Longueur
    Tip Color
    Packed
  • LTS compatible, extended length
    20 µl
    +
    +
    49.45 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $170.00
    $170.00
    Case of 4800
    Case of 4800 - $850.00
    $850.00
    Pack of 960
       $170.00
    Case of 4800
       $850.00
     : LTFER-20-R-S
      Sustainable Packaging|Low Manufacturing Impact
    Specifications
    Cat. No.
    77395-598
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Environmental certification
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Sustainable Packaging|Low Manufacturing Impact
  • LTS compatible
    200 µl
    +
    +
    50.29 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $170.00
    $170.00
    Case of 4800
    Case of 4800 - $850.00
    $850.00
    Pack of 960
       $170.00
    Case of 4800
       $850.00
     : LTFER-200-R-S
      Sustainable Packaging|Low Manufacturing Impact
    Specifications
    Cat. No.
    77395-600
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Environmental certification
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Sustainable Packaging|Low Manufacturing Impact
  • LTS compatible, extended length
    1000 µl
    +
    +
    98.58 mm
    Clear
    10 racks, 96 each
    Pack of 960
    Pack of 960 - $178.00
    $178.00
    Case of 4800
    Case of 4800 - $890.00
    $890.00
    Pack of 960
       $178.00
    Case of 4800
       $890.00
     : LTFER-1000-R-S
      Sustainable Packaging|Low Manufacturing Impact
    Specifications
    Cat. No.
    77395-602
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Environmental certification
    Compared to conventional plastic racked pipet tips, reduces plastic per case by 70%.
    Produced in ISO 14001 Certified facility.
      Sustainable Packaging|Low Manufacturing Impact