Order Entry
Canada
ContactUsLinkComponent
Anti-CD86 (B7-2) Rat Monoclonal Antibody [clone: GL-1] (FITC)
Catalogue # MSPP-100-1626
Fournisseur:  STEMCELL Technologies
undefined
Anti-CD86 (B7-2) Rat Monoclonal Antibody [clone: GL-1] (FITC)
Catalogue # MSPP-100-1626
Fournisseur:  STEMCELL Technologies
Supplier Number:  100-1626

Spécifications

  • Antigen name:
    CD86 (B7-2)
  • Clonality:
    Monoclonal
  • Clone:
    GL-1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Size:
    100 µg
  • Gene ID:
    12524
  • Cat. no.:
    MSPP-100-1626

Spécifications

À propos de cet article

Rat monoclonal antibody against mouse CD86 (B7-2), FITC-conjugated.

  • B cells
  • Dendritic cells
  • Macrophages

This monoclonal (clone GL-1) antibody reacts with mouse cluster of differentiation 86 (CD86; also known as B7-2), an ~80 kDa immunoglobulin superfamily member and a type I transmembrane glycoprotein. Both CD80 and CD86 are ligands of the T cell surface proteins CD28 and CD152 (CTLA-4); however, CD86 is expressed earlier in the immune response compared to CD80. CD86 is also involved in immunoglobulin class-switching and triggering of natural killer cell-mediated cytotoxicity. CD86 binds to CD28 to transduce co-stimulatory signals for T cell activation, proliferation, and cytokine production. CD86 can also bind to CTLA-4 with a 20- to 100-fold higher affinity than CD28 and deliver an inhibitory signal to T cells as well as down regulate the immune response. CD86 has been used as a phenotypic marker for differentiating cells. This antibody can be used as a marker to assess classically activated M1 murine macrophages. Use this recombinant monoclonal antibody in applications requiring detection of CD86, such as flow cytometry (FC).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD86 (B7-2)
Clonality: Monoclonal
Clone: GL-1
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse