Order Entry
Canada
ContactUsLinkComponent
WarmStart® Bst 2.0 DNA Polymerase, Glycerol-free
Catalog # CA77532-244
New Product
CAS Number:  
WarmStart® Bst 2.0 DNA Polymerase, Glycerol-free
Catalog # CA77532-244
Supplier Number:  M0402L
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    120000 units/ml
  • Description:
    Bst 2.0 DNA polymerase, glycerol-free
  • Size:
    8000 units
  • Cat. no.:
    CA77532-244

Specifications

About this item

Bst 2.0 WarmStart DNA polymerase (glycerol-free) is formulated without glycerol to support lyophilization, incorporation into microfluidic devices, and enable quick adoption into automation workflows.

  • Glycerol-free format to support lyophilization and automation workflows
  • Consistent amplification performance with the convenience of room temperature setup
  • WarmStart technology eliminates off-target amplification and offers increased reaction efficiency
  • Higher salt tolerance and thermostability compared to wild-type Bst DNA polymerase, large fragment
  • Optimal reaction performance from 60 to 72 °C

Bst 2.0 WarmStart® DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA polymerase I, large fragment (Bst DNA polymerase, large fragment) with a reversibly-bound aptamer which inhibits polymerase activity at temperatures below 45 °C. At temperatures above 45 °C, the aptamer rapidly releases from the enzyme with no special activation step needed to activate the polymerase. Bst 2.0 WarmStart® DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand-displacement activity but lacks 5' to 3' exonuclease activity. It displays improved amplification speed, yield, salt tolerance, and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.