Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:ErythroClear™ Magnet
- Cat. no.:MSPP-01737
Specifications
About this item
Perform total nucleic acid extraction from a variety of sample types using the ErythroClear™ magnet.
- Magnet for column-free immunomagnetic separation
- Process up to 16 standard polypropylene microcentrifuge tubes at a time
Perform total nucleic acid extraction from a variety of sample types, including whole blood and cell suspensions (PBMCs, hPSCs, cultured cells), using the ErythroClear™ magnet in combination with EasySep™ reagents. Use the ErythroClear™ magnet to process up to 16 standard polypropylene microcentrifuge tubes at a time (8 samples per side).