SteriWare Multi Grip Tweezers
New Product
Supplier: SAMPLING SYSTEMS LTD
Jump to:
À propos de cet article
These SteriWare multi grip tweezers operate like conventional tweezers, they have an additional grip feature enabling the easy and secure handling of small vials and containers.
- Made from food grade PS
- Individually bagged to prevent cross contamination
- 100% manufactured and packed in a cleanroom
- Choice of sterile and non sterile
4 Options Available Below
Product Details & Documents
These SteriWare multi grip tweezers operate like conventional tweezers, they have an additional grip feature enabling the easy and secure handling of small vials and containers.
- Made from food grade PS
- Individually bagged to prevent cross contamination
- 100% manufactured and packed in a cleanroom
- Choice of sterile and non sterile
Documents
Product Family Options
Sterile
Informations sur le produit
- DésignationMatièreLongueurAvailabilityPrice
- Supplier Number: 8270A-300SSpécifications:New ProductNew Product
- Supplier Number: 8270A-200SSpécifications:New ProductNew Product
Non sterile
Informations sur le produit
- DésignationMatièreLongueurAvailabilityPrice
- Supplier Number: 8270A-300Spécifications:New ProductNew Product
- Supplier Number: 8270A-200Spécifications:New ProductNew Product