SteriWare Micro-Spatula Samplers | Avantor
SteriWare Micro-Spatula Samplers
New Product
SteriWare Micro-Spatula Samplers
New Product
Certificates
SteriWare Micro-Spatula Samplers
SteriWare Micro-Spatula Samplers
3 Options Available Below
Product Details & Documents
Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.
  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use
Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Volume
    Availability
    Price
  • Sterile micro-spatula, 13 mm blade width
    PP
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $230.42
    $230.42
    Case of 100
       $230.42
    Supplier Number: 8015B-170S
    Spécifications:
    New Product
    New Product
  • Sterile micro-spatula, 6 mm blade width
    PP
    0.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $230.42
    $230.42
    Case of 100
       $230.42
    Supplier Number: 8015NB-170S
    Spécifications:
    New Product
    New Product
  • Sterile micro-spatula, 4 mm blade width
    PP
    0.3 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $230.42
    $230.42
    Case of 100
       $230.42
    Supplier Number: 8015VB-170S
    Spécifications:
    New Product