Pre-Printed ID Tapes | Avantor
Pre-Printed ID Tapes
New Product
Pre-Printed ID Tapes
New Product
Certificates
Pre-Printed ID Tapes
Pre-Printed ID Tapes
4 Options Available Below
Product Details & Documents
ID tape helps to prevent handling errors by allowing containers, pallets, bags and so on to be easily labelled with highly visible messaging. Operators can quickly identify the status of a container to prevent it from being used in error.
  • Use with standard tape dispensers
  • Medium strength adhesion
ID tape is an ideal way of labelling large or small items. These tape rolls come pre-printed with 'Quarantine', 'Sampled', 'Released' or 'Rejected'.
Packaging: 10 rolls per box.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Largeur
    Availability
    Price
  • ID tape, printed with 'Quarantine'
    White/red
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $135.99
    $135.99
    Case of 10
       $135.99
    Supplier Number: 8221A-01
    Spécifications:
    New Product
    New Product
  • ID tape, printed with 'Sampled'
    White/red
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $135.99
    $135.99
    Case of 10
       $135.99
    Supplier Number: 8221A-02
    Spécifications:
    New Product
    New Product
  • ID tape, printed with 'Released'
    White/red
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $135.99
    $135.99
    Case of 10
       $135.99
    Supplier Number: 8221A-03
    Spécifications:
    New Product
    New Product
  • ID tape, printed with 'Rejected'
    White/red
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $135.99
    $135.99
    Case of 10
       $135.99
    Supplier Number: 8221A-04
    Spécifications:
    New Product
    New Product