SteriWare® V Spatulas for Powder Sampling
About this item
These V spatulas are ideal for sampling food, chemical and pharmaceuticals. These sampling tool are ideal for powders, creams and pastes. To ensure cleanliness they are manufactured and then individually packed in a class 7 cleanroom (10000).
- Robust PS construction
- Available in narrow 10 mm width and standard 20 mm width
- Both non sterile and sterile available
- Blade lengths from 100 to 250 mm
Ideal for goods in quality control or just general sampling. The two widths and three lengths give ideal combination and choice.
: Both EU and FDA compliant.
: Individually bagged.
12 Options Available Below
Product Details & Documents
These V spatulas are ideal for sampling food, chemical and pharmaceuticals. These sampling tool are ideal for powders, creams and pastes. To ensure cleanliness they are manufactured and then individually packed in a class 7 cleanroom (10000).
- Robust PS construction
- Available in narrow 10 mm width and standard 20 mm width
- Both non sterile and sterile available
- Blade lengths from 100 to 250 mm
Ideal for goods in quality control or just general sampling. The two widths and three lengths give ideal combination and choice.
: Both EU and FDA compliant.
: Individually bagged.
Product Family Options
Product Information
- ColorLongueurBlade WidthSterilityAvailabilityPrice
- Supplier Number: 8440N-100Specifications:New ProductNew Product
- Supplier Number: 8440N-100SSpecifications:New ProductNew Product
- Supplier Number: 8440N-150Specifications:New ProductNew Product
- Supplier Number: 8440N-150SSpecifications:New ProductNew Product
- Supplier Number: 8440N-250Specifications:New ProductNew Product
- Supplier Number: 8440N-250SSpecifications:New ProductNew Product
- Supplier Number: 8440A-100Specifications:New ProductNew Product
- Supplier Number: 8440A-100SSpecifications:New ProductNew Product
- Supplier Number: 8440A-150Specifications:New ProductNew Product
- Supplier Number: 8440A-150SSpecifications:New ProductNew Product
- Supplier Number: 8440A-250Specifications:New ProductNew Product
- Supplier Number: 8440A-250SSpecifications:New ProductNew Product