Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human/Mouse Recombinant TGF-beta 3 | Avantor

Human/Mouse Recombinant TGF-beta 3

1000 µg
  MSPP-781311997
Marketsource
Fournisseur #:  78131.2
Human/Mouse Recombinant TGF-beta 3
  MSPP-781311997
Marketsource
 :  
Human/Mouse Recombinant TGF-beta 3
Human/Mouse Recombinant TGF-beta 3

1000 µg

 :  
 
$11,036.92
   
Each(1/Itm/Bx)
La quantité minimale de commande est de 1
 

Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes. TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.). TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.). TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

  • Taille:
    1000 µg
  • Cat. No.:
    MSPP-781311997
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TGF-beta 3
  • Purity:
    ≥95%
  • Species:
    Human|Mouse

 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.