Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Size:1 unit
- Protein/peptide name:HIV-1 (B Gag) Peptide Pool
- Purity:Average 70%
- Cat. no.:MSPP-100-1385
Spécifications
À propos de cet article
Stimulating T cells with HIV-1 (B Gag) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HIV-1 (B Gag) Peptide Pool is a lyophilized mixture of 123 peptides from the gag polyprotein of human immunodeficiency virus 1 (HIV-1). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 500 on gag polyprotein. The HIV-1 gag polyprotein is a key mediator of viral particle assembly (Campbell and Rein; Dong et al.), and together with the endosomal sorting complexes required for transport (ESCRT), results in budding during virion release (Carlson et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.