Cleanroom Sticky Notes
Cleanroom Sticky Notes
Cleanroom Sticky Notes
Cleanroom Sticky Notes
Class 10 cleanroom sticky notes include 100 latex sheets per pad and are designed for use in critical environments where low particulation and ESD's are required.
  • 28# latex paper
  • 3×3" note pads
  • 24 pads per case
The latex sheets are smudge-resistant and the adhesive used for binding the sheets does not emit ESD's or carry particulates when a sheet is removed.
Product Family Options
  • Désignation
    W×L
    Color
  • Class 10 cleanroom sticky notes
    7.6×7.6 cm (3×3")
    Bright yellow
    Case of 24
    Case of 24 - $260.24
    $260.24
    Case of 24
       $260.24
     : POST-IT-CAN
  • Class 10 cleanroom sticky notes
    7.6×7.6 cm (3×3")
    Green
    Case of 24
    Case of 24 - $260.24
    $260.24
    Case of 24
       $260.24
     : POST-IT-GRE