Science Take-Out® Disappearing Bees
Why are the bees dying?
- Available as a single kit with materials for one student or group
- Unassembled ten-pack for additional savings
- No additional equipment needed
Bees are dying! Explore how the loss of bees will affect your food supply. Use simulated lab tests and data from scientific research to identify possible reasons for the decline of honey bee populations. Explore actions that could be taken to protect insect pollinators.
Frequently Bought Together