Cleanroom Storage Rack, Stainless Steel, Bandy
Catalogue # 98000-584
Fournisseur: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Finish:No. 4
- Width:12"
- Description:Cleanroom storage rack
- Height:67"
- Length:27"
- Material:Stainless steel 304
- Cat. no.:98000-584
- Supplier No.:DISP60001
Spécifications
À propos de cet article
Twelve-compartment rack is ideal for storage of disposable cleanroom items such as gloves, booties, or bouffant caps.
- All surface perforated
- Perforations allow appropriate air flow
- Adjustable leveling feet
Constructed of 16-gauge, Type 304 stainless steel with a No. 4 finish.