SC3 Super Compact Temperature Recorders, Dickson®
À propos de cet article
Reusable and compact recorders are built with a rugged case to withstand the rigors of transportation and storage applications
Economical recorders feature easy user calibration and switchable control of temperature, °C/°F, and time, 24 hours/7 days. Designed with specialized slots for unit security. Optional tamper seals can be used to ensure the integrity of the chart. Units can be wall-mounted, suspended, or used free-standing, and work with charts that have a diameter of 75mm (3").
: Supplied with one AA battery, pen, and instruction manual.
9 Options Available Below
Product Details & Documents
Reusable and compact recorders are built with a rugged case to withstand the rigors of transportation and storage applications
Economical recorders feature easy user calibration and switchable control of temperature, °C/°F, and time, 24 hours/7 days. Designed with specialized slots for unit security. Optional tamper seals can be used to ensure the integrity of the chart. Units can be wall-mounted, suspended, or used free-standing, and work with charts that have a diameter of 75mm (3").
: Supplied with one AA battery, pen, and instruction manual.
Specifications for Product Family
Spécifications
- Catalog No:
- 14905-004
- 14905-006
- 14905-008
- 14905-010
- 14905-012
- 61280-024
- 61280-028
- 61280-032
- 61280-034
- 61280-040
- 61280-044
- 61280-490
- 61280-540
- 61280-020
- 61280-547
- Accuracy:
- ±1°C (±2°F)
- ±1°C (±2°F)
- ±1°C (±2°F)
- ±1°C (±2°F)
- ±1°C (±2°F)
—
—
—
—
—
—- —
- —
- —
- —
- Dimensions:
- 9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")
- 9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")
- 9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")
- 9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")
- 9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")
—
—
—
—
—
—- —
- —
- —
- —
- Weight:
- 227 g (8 oz.)
- 227 g (8 oz.)
- 227 g (8 oz.)
- 227 g (8 oz.)
- 227 g (8 oz.)
—
—
—
—
—
—- —
- —
- —
- —
Spécifications
Product Family Options
Chart Recorders
Informations sur le produit
- Plage de températureAvailabilityPrice
- Supplier Number: SC367Spécifications:Small Business Enterprise
More
Specifications
Cat. No.14905-004Accuracy±1°C (±2°F)Dimensions9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")Weight227 g (8 oz.)Small Business Enterprise - Supplier Number: SC377Spécifications:Small Business Enterprise
More
Specifications
Cat. No.14905-006Accuracy±1°C (±2°F)Dimensions9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")Weight227 g (8 oz.)Small Business Enterprise - Supplier Number: SC386Spécifications:Small Business Enterprise
More
Specifications
Cat. No.14905-008Accuracy±1°C (±2°F)Dimensions9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")Weight227 g (8 oz.)Small Business Enterprise - Supplier Number: SC387Spécifications:Small Business Enterprise
More
Specifications
Cat. No.14905-010Accuracy±1°C (±2°F)Dimensions9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")Weight227 g (8 oz.)Small Business Enterprise - 25...50 °C (76...122 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.14905-012Accuracy±1°C (±2°F)Dimensions9.3L x 9.3W x 5.7H cm (311/16 x 311/16 x 23/16")Weight227 g (8 oz.)Small Business Enterprise
Temperature Charts
Informations sur le produit
- Plage de températureAvailabilityPrice
- Supplier Number: C314Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-024Accuracy
—Dimensions
—Weight
—Small Business Enterprise - 25...50 °C (76...122 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-028Accuracy
—Dimensions
—Weight
—Small Business Enterprise - –25...0 °C (–14...32 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-032Accuracy
—Dimensions
—Weight
—Small Business Enterprise - Supplier Number: C310Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-034Accuracy
—Dimensions
—Weight
—Small Business Enterprise - Supplier Number: C312Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-040Accuracy
—Dimensions
—Weight
—Small Business Enterprise - –15...10 °C (4...50 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-044Accuracy
—Dimensions
—Weight
—Small Business Enterprise - 10...35 °C (50...96 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-490Accuracy—Dimensions—Weight—Small Business Enterprise - –5...20 °C (22...68 °F)Discontinued-Supplier Number: -Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-540Accuracy—Dimensions—Weight—Small Business Enterprise - Supplier Number: C316Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-020Accuracy—Dimensions—Weight—Small Business Enterprise - –5...20 °C (22...68 °F)Case of 60$188.58Each$188.58Supplier Number: C318Spécifications:Small Business Enterprise
More
Specifications
Cat. No.61280-547Accuracy—Dimensions—Weight—Small Business Enterprise