Drop-In Sinks, Durcon Laboratory Tops
: Durcon
One-piece sinks are molded of epoxy resin with coved corners and a bottom sloped to the sink outlet.
- Sink outlet is required for use
ADA-compliant sinks are also available.
: Sink outlet and overflow are offered separately (see DU-CS4R series).
: Stoppers are supplied with this product.
Frequently Bought Together