À propos de cet article
Elevate your ID card printing with the Evolis Primacy 2 Ribbons, specifically engineered for compatibility with the Evolis Primacy 2 printer. This high-quality, full-color YMCKO ribbon delivers professional-grade, vibrant, and long-lasting prints for your ID cards, badges, and access control cards.
- Designed for seamless integration with the Evolis Primacy 2 printer, ensuring optimal performance and reliability
- This ribbon offers cost-effective printing solutions for your ID card projects
- The K panel is a black resin panel, typically used for printing text and barcodes
- This ribbon is specially designed to guarantee optimum operation of your badge printer without risk of damage
- Design allows for quick and hassle-free replacement, minimizing downtime and ensuring a smooth printing process
Expertly designed for seamless integration, guaranteeing optimal performance and reliability. Produce stunning, full-color ID cards with sharp images, clear text, and vivid colors, utilizing the advanced dye-sublimation YMCKO-K technology in the ribbon.
With an impressive capacity of 200 prints per ribbon, this ribbon offers cost-effective printing solutions for your ID card projects. The ribbon's user-friendly design allows for quick and hassle-free replacement, minimizing downtime and ensuring a smooth printing process.
Ribbon consists of yellow (Y), magenta (M) and cyan (C) panels for printing a full spectrum of colors by combining the colors using varying degrees of heat.