ContactUsLinkComponent
SP Bel-Art Poxygrid® Microliter Pipettor Racks, Bel-Art Products, a part of SP
SP Bel-Art Poxygrid® Microliter Pipettor Racks, Bel-Art Products, a part of SP
Catalog # 53577-145
undefined
SP Bel-Art Poxygrid® Microliter Pipettor Racks, Bel-Art Products, a part of SP
Catalog # 53577-145
Supplier Number:  H18962-0000
Specifications
  • Description:
    For Finnpipette®, Eppendorf®, and Other Popular Brand Single-Channel Pipettors
  • Dimensions:
    22.9W x 12.7D x 17.1H cm (9 x 5 x 63/4")
  • Cat. no.:
    53577-145
  • Supplier No.:
    189620000
Specifications
About this item
Store pipettors while protecting pipettor tips in these epoxy-coated aluminum racks that provide quick, easy access for more efficient work.
  • Epoxy-coated aluminum rack construction
  • Hold a range of pipettors
  • Suitable for wall mounting or free standing
Able to hold up to 10 round-shaped pipettors such as Finnpipette®, Socorex®, Eppendorf® and other popular brands in two stepped rows of 5 each. This rack also has end holes for the Pipet-Aid® pipette filler. A second lower support row has smaller holes that accept the tapered ends of the pipettors to keep them vertical.