Wall Mounted Needle Valve, WaterSaver Faucet
White, epoxy-coated, single straight pattern laboratory needle valve.
Brass construction with stainless steel needle and seat. Needle and seat are removable/replaceable for infinite service life. Black, nylon, hooded handle with color-coded index button. Mounting shank included. Rated for use at pressures up to 10.3 bar (150 psi).
: Please specify service when ordering.
Frequently Bought Together