ContactUsLinkComponent
Magicard® YMCKOK Color Ribbon
Catalogue # 77531-630
Fournisseur:  Janitorial Supplies
New Product
Magicard® YMCKOK Color Ribbon
Catalogue # 77531-630
Fournisseur:  Janitorial Supplies
Supplier Number:  SRXMA250YMCKOK
New Product
Spécifications
  • For Use With:
    Magicard® Enduro 3E, Pronto, Rio Pro, Rio Pro 360, Rio Pro Secure
  • Color:
    Black, cyan, magenta, yellow
  • Description:
    Magicard® YMCKOK color ribbon, 3.13×5.13×2.38" (W×H×D), 0.350 lbs.
  • Cat. no.:
    77531-630
Spécifications
À propos de cet article
Magicard® YMCKOK ribbons are ideal for dual-sided card printing with YMC, the first K panel, and O panel printing on the front side of the card and the second K panel printing on the back side of the card.
  • Full color six-panel YMCKOK dye film for dual-sided printing
  • Print full-color images on the front and one-color text or barcodes on the back
  • Features drop-in dye film loading which provides hassle-free installation
  • Estimated print yield of 250 individual single-sided card prints
Magicard ® consumables are specially designed, tested and approved for Magicard® printers to ensure high quality images and text. Genuine Magicard® consumables offer the best performance and extended lifespan of your card printer.