- Color:Black, cyan, magenta, yellow, topcoat protective layer
- Description:Datacard® Entrust YMCKT 2-sided full color ribbon kit, 3.20×3.00×5.30" (W×H×D), 0.475 lbs.
- Cat. no.:77531-614
Entrust Datacard YMCKT-KT full-color ribbon includes full spectrum color overlay to protect against fading colors. This printer ribbon produces rich, full-color images and one-color barcodes or text on cards. For single-sided card printing or dual-sided printing.
- Fits the ENTRUST DS2 and DS3 card printers
- Includes two topcoat panels for enhanced fade resistance and card protection; both can be applied to front of card for extended card life
Entrust YMCKT-KT ribbons are used for printing a combination of full-color images and one-color text or barcodes. YMCKT-KT ribbons consist of yellow (Y), magenta (M) and cyan (C) panels for printing a full spectrum of colors by combining colors using varying degrees of heat.
The two K panels are black resin, typically using for printing text and barcodes and the two T panels are thin, clear protective overlays. Each print kit includes one ribbon, one isopropanol cleaning card and one adhesive cleaning roller.
A 7-panel YMCKT-KT print ribbon that is designed for double sided printing and will produce full-color, photo-quality images on the front; and black resin printing of crisp text, logos, and barcodes on the back of your cards