Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Catalogue # 60004-228
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware Heat Transfer Block for Minivials, Microscale
- Cat. no.:60004-228
- Supplier No.:H200001
Spécifications
À propos de cet article
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
- Aluminum heat transfer block
- Hold three sizes of vials
Block has two holes for 3 ml and 5 ml heavy wall reaction vials, four holes for Craig recrystallization tubes, and one hole for a standard thermometer.