Swinging/Fixed Gooseneck Coupling, WaterSaver Faucet
VFT-0520-00
- Description:Swinging/Fixed gooseneck coupling
- Cat. no.:VFT-0520-00
- Supplier No.:B0356COMP
This coupling is designed for use with all standard goosenecks and it replaces washer in a fixed gooseneck faucet, converting it to a swing faucet.
- Useful for double tub sinks
- With 1 cm (3/8") IPS male threads