Nalgene® Rectangular Carboys with Spigot and Handle, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Excellent for storage and dispensing of buffers, acids, and bases.
- Tough, rigid translucent carboy features molded-in graduations
- Wide mouth allows easy, efficient cleaning
- Built-in shoulder loops have sturdy handle attached
- Leakproof
Ordering information: Supplied with carboy, polypropylene spigot, and 100-415 screw cap. For replacement caps, see cap 16219-106 in 16219-030 series.
Frequently Bought Together